Learn More
KN-62, MedChemExpress
MedChemExpress KN-62 is a selective and reversible inhibitor of calmodulin-dependent protein kinase II (CaMK-II) with a Ki of 0.9 μM for rat brain CaMK-II. KN-62 directly binds to the calmodulin binding site of CaMK-II. KN-62 displays noncompetitive antagonism at P2X7 receptors in HEK293 cells, with an IC50 value of approximately 15 nM.
Brand:  Medchem Express HY-13290
Encompass_Preferred
 View more versions of this product
Product Code. 16425628
  • 209.25 € / 10mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
KN-62 is a selective and reversible inhibitor of calmodulin-dependent protein kinase II (CaMK-II) with a Ki of 0.9 μM for rat brain CaMK-II. KN-62 directly binds to the calmodulin binding site of CaMK-II. KN-62 displays noncompetitive antagonism at P2X7 receptors in HEK293 cells, with an IC50 value of approximately 15 nM.
Specifications
Specifications
KN-62
Yellow
C38H35N5O6S2
DMSO : ≥ 100 mg/mL (138.53 mM)
721.84
98.27%
Powder
127191-97-3
Research
10 mg
O=C(N1CCN(C2=CC=CC=C2)CC1)[C@H](CC3=CC=C(C=C3)OS(=O)(C4=CC=CC5=C4C=CN=C5)=O)N(C)S(=O)(C6=CC=CC7=C6C=CN=C7)=O
721.84
Research
Cancer-Kinase/protease
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
KN-62, MedChemExpress