Learn More
KN-62, MedChemExpress
MedChemExpress KN-62 is a selective and reversible inhibitor of calmodulin-dependent protein kinase II (CaMK-II) with a Ki of 0.9 μM for rat brain CaMK-II. KN-62 directly binds to the calmodulin binding site of CaMK-II. KN-62 displays noncompetitive antagonism at P2X7 receptors in HEK293 cells, with an IC50 value of approximately 15 nM.

122.85 € - 843.75 €
Specifications
Chemical Name or Material | KN-62 |
---|---|
CAS | 127191-97-3 |
Color | Yellow |
Purity Grade Notes | Research |
Molecular Formula | C38H35N5O6S2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478317
![]() |
Medchem Express
HY-13290 |
5 mg |
122.85 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16425628
![]() |
Medchem Express
HY-13290 |
10 mg |
209.25 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411797
![]() |
Medchem Express
HY-13290 |
50 mg |
843.75 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KN-62 is a selective and reversible inhibitor of calmodulin-dependent protein kinase II (CaMK-II) with a Ki of 0.9 μM for rat brain CaMK-II. KN-62 directly binds to the calmodulin binding site of CaMK-II. KN-62 displays noncompetitive antagonism at P2X7 receptors in HEK293 cells, with an IC50 value of approximately 15 nM.Specifications
KN-62 | |
Yellow | |
C38H35N5O6S2 | |
O=C(N1CCN(C2=CC=CC=C2)CC1)[C@H](CC3=CC=C(C=C3)OS(=O)(C4=CC=CC5=C4C=CN=C5)=O)N(C)S(=O)(C6=CC=CC7=C6C=CN=C7)=O | |
721.84 | |
Research | |
Cancer-Kinase/protease |
127191-97-3 | |
Research | |
DMSO : ≥ 100 mg/mL (138.53 mM) | |
721.84 | |
98.27% | |
Powder |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.