Learn More
Greiner Bio-One Polypropylene Microcentrifuge Tubes
For use in safe and long term storage. Greiner Bio-One Polypropylene Microcentrifuge Tubes offer a leak-free performance and minimal non-specific binding.
109.00 € - 600.00 €
Products 4
Product Code Brand Includes Closure Color Price Quantity & Availability  
Product Code Brand Includes Closure Color Price Quantity & Availability  
10687954
View Documents
Greiner Bio-One
716201
N/A Natural
321.43 €
Pack of 5000
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15975351
View Documents
Greiner Bio-One
717201
Support skirt Natural
392.86 €
Pack of 5000
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
10534991
View Documents
Greiner Bio-One
717201
Support skirt Natural N/A N/A N/A
10111852
View Documents
Greiner Bio-One
717261
Support skirt White
600.00 €
Pack of 3000
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • chemical and temperature resistance
  • clarity coupled to a wide thermal range
  • May be centrifuged to 20,000g
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One Polypropylene Microcentrifuge Tubes