Learn More
Thermo Scientific™ Orion™ 2230XP Silica Analyzer Reagents and Standards
The reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer.
499.00 €
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15914267
View Documents
Thermo Scientific™
2230RE
Reagents 1,2 and 3
499.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Ensure you have the reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer. Reagents kit contains all three reagents required for operation, and calibration standards available in individual bottles. For use with the Orion 2230XP Silica Analyzer.
Warranty: Out of box
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contains : sodium metasilicatenonahydrate
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Orion™ 2230XP Silica Analyzer Reagents and Standards